Learn More
Medchemexpress LLC 2-Butenedioic acid (2E)-, 1,4-diethyl ester | 623-91-6 | 98.97% | 172.18 | 50 G
Supplier: Medchemexpress LLC HYW01005650G
Diethyl fumarate is an analytical standard intended for research and analytical applications. It is a decomposition product of Malathion 9 (a pesticide) and a reactant in the Diels-Alder reaction. Diethyl fumarate is a reputed skin irritant and can cause non-immunologic contact urticaria. It can also hybridize with Curcumin to exert neuroprotective effects. The compound is of analytical standard grade and is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
- Analytical standard for research and analytical applications
- Used in qualitative, quantitative, and methodological research experiments
- Suitable for HPLC, GC, and MS applications
- Can exert neuroprotective effects when hybridized with Curcumin
- Reactant in Diels-Alder reaction