Learn More
Med Vet International LASER/10MM SM SPOT CERAMIC TIP
Supplier: Med Vet International JOR941B
For use with the AccuVet LX-20 CO2 Laser System JorVet offers these commonly needed parts for the operation of the CO2 Laser These parts are warranted to be identical in quality to any factory part All laser tips are compatible with Luxar LXH-ST LXH-TL LXHA105 and LX-AH Handpieces Non-sterile