Learn More
Med Vet International BODYSUIT/VETMED CARE/SM/MALE
Supplier:  Med Vet International JOR1055BM
Encompass_Preferred
This new line of protective body wear has been used extensively by veterinarians throughout Europe A wide range of applications has evolved literally from head to tail on a whole host of different species Traditional methods of wound protection such as E collars and adhesive tapes have several known disadvantages The VetMedCare protective body wear offers a great alternative to wound protection Body suits are easy to handle and protect sensitive areas from dirt scratching and licking Thoughtful design provides front and back leg openings that have special flat and stretch seams which help avoid limb irritations or abrasions The belly is fully smooth as the seam is along the back Anatomical differences require consideration of both male and female designs Material/Sizes Very elastic stretches in all directions for a comfortable fit Wash and wear material
Catalog No. 50-004-95607