Learn More
Gold Biotechnology Inc Bluo Gal 250 mg
SDP
Supplier:  Gold Biotechnology Inc B673250
Encompass_Preferred
Bluo-Gal is used as an alternative to X-Gal and produces a darker blue color Bluo-Gal is hydrolyzed by -galactosidase to produce a blue precipitate This precipitate is insoluble in alcohol and xylenes thus making it suitable for immunoblotting and immunocytochemical assays Bacterial colonies containing active -galactosidase produce blue colonies when grown on plates containing Bluo-Gal
Catalog No. 50-005-62081