Learn More
Anton Paar Cora 5001 direct raman spectrometer with sample compartment, wavelength 785nm
Supplier: Anton Paar 225834
Single Wavelength or Dual Wavelength.Available wavelengths:532nm785nm1064nmBattery operation option