Learn More
Ace Glass, Inc. 5000ML CYLINDER 5000ML CYLINDER
SDP
Supplier:  Ace Glass, Inc. 622805
Encompass_Preferred
5000mL cylindrical graduated beaker for batch operations and mixing large volumes of measured liquids.
Catalog No. 50-978-663