Learn More
Chemglass Life Sciences 500mL Erlenmeyer Flask, 38-430 GPI Screw Thread
SDP
Supplier:  Chemglass Life Sciences CG154304
Encompass_Preferred
500mL Erlenmeyer Flask, 38-430 GPI Screw Thread. Erlenmeyer flask with screw thread finish. Supplied complete with white rubber lined black phenolic cap. Long skirted caps can be loosened without falling off while shaking flask.
Catalog No. CG154304