Learn More
Hach Company 500mL Flask-Volumet-Polypro
Supplier:  Hach Company 1406049
Encompass
Polypropylene flask and screw cap Durable with excellent chemical resistance with the same accuracy as glass Permanent color graduation line with marking spot Calibrated to contain/deliver at 20 C The 200/203 capacity flask has dual marks useful in DO determination via Winkler Titration
Catalog No. 50-002-57137