Learn More
Glas Col LLC Heating Mantle
SDP
Supplier:  Glas Col LLC 100A O406/DUP
Encompass_Preferred
Fabric mantle for 500ml spherical flask, to 400 degrees C maximum internal operating temperature, 270W, 115V CSA
Catalog No. NC1920315