Learn More
ACROBiosystems Recombinant Protein;Human FGF basic Protein, Tag Free;E.coli;50UG;BFF,FGF2,FGF-2,BFGF,FGFB,FGFβ,FGF-β,FGF basic,HBGF-2,HBGF2
Supplier:  ACROBiosystems BFFH411750UG
Encompass_Preferred
Recombinant Protein;Human FGF basic Protein, Tag Free;50UG;Human FGF basic, Tag Free (BFF-H4117) is expressed from E.coli cells. It contains AA Pro 143 - Ser 288 (Accession # NP_001997).;This protein carries no "tag". The protein has a calculated MW of 16.5 kDa. The protein migrates as 17 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 2μg/mL (100 μL/well) can bind Human Glypican 3, Fc Tag (Cat. No. GP3-H5258) with a linear range of 0.48-7.8 ng/mL (QC tested).
Catalog No. 50-101-7274