Learn More
ACROBiosystems Recombinant Protein;Cynomolgus CD3 epsilon & CD3 delta Heterodimer Protein, His Tag & Flag Tag;HEK293;100UG;CDD,CD3E & CD3D,FLJ18683,T3E,TCRE,CD3E,CD3-epsilon,CD3D,CD3-DELTA,T3D,CD3E&CD3D,CD3 delta & CD3 epsilon,CD3 delta / epsilon
Supplier:  ACROBiosystems CDDC52W4100UG
Encompass_Preferred
Recombinant Protein;Cynomolgus CD3 epsilon & CD3 delta Heterodimer Protein, His Tag & Flag Tag;100UG;Recombinant Cynomolgus CD3E & CD3D heterodimer protein (Cynomolgus CD3E & CD3D heterodimer) Gln 22 - Asp 117 (CD3E,Q95LI5) & Phe 22 - Ala 105 (CD3D,Q95LI8) was produced in human 293 cells (HEK293) at ACROBiosystems.;The heterodimer Cynomolgus CD3E & CD3D heterodimer is produced by co-expression of CD3E and CD3D, has a calculated MW of 15.5 kDa (CD3E) and 14.5 kDa (CD3D). Subunit CD3E is fused with his tag at the C-terminus and subunit CD3D is fused with flag tag at the C-terminus. The predicted N-terminus is Gln 22 (CD3E) and Phe 22 (CD3D). The reducing (R) heterodimer protein migrates as 19-22 kDa and 25-40 kDa due to glyosylation.;Immobilized SP34-2 at 5 μg/mL (100 μL/well) can bind Cynomolgus CD3E & CD3D, His Tag, Flag Tag (Cat. No. CDD-C52W4) with a linear range of 0.6-5 ng/mL (Routinely tested).
Catalog No. 50-101-7483
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.