Learn More
ACROBiosystems Recombinant Protein;Human IgG4 Fc Protein, Tag Free;HEK293;200UG;IG4,IGHG2,IgG2 Fc,IgG2A Fc,IgG1 Fc,IgG2,IgG3,IgG4,IgG2a,IgG Fc,IgG1,IgG2a Fc,IgG4 H Chain,IGHG4,IgG4 Fc
Supplier:  ACROBiosystems IG4H5205200UG
Encompass_Preferred
Recombinant Protein;Human IgG4 Fc Protein, Tag Free;200UG;Human IgG Fc, Tag Free (IG4-H5205) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 327 (Accession # P01861).;This protein carries no "tag". The protein has a calculated MW of 25.8 kDa. As a result of glycosylation, the protein migrates as 33-35 kDa under reducing (R) condition, and 33 kDa and 46-50 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-101-8022