Learn More
New England Biolabs, Inc. NEBNext® Ultra™ II Ligation Module – 96 reactions
SDP
Supplier:  New England Biolabs, Inc. E7595L
Encompass
This module is part of the Ultra II workflow, and is optimized for use with the NEBNext Ultra II End Repair/dA-Tailing Module, for Illumina-compatible library construction. The NEBNext Ultra II Ligation Module is optimized for use with the NEBNext Ultra II End Repair/dA-Tailing Module or the NEBNext Ultra II FS DNA Module. This module is also compatible with some Oxford Nanopore Technologies workflows.
  • Optimized to ligate DNA adaptors for Illumina sequencing to end-repaired, dA-tailed DNA fragments
  • Compatible with some Oxford Nanopore Technologies workflows
Catalog No. 50-105-0630