Learn More
ACROBiosystems Recombinant Protein;Mouse CTLA-4 / CD152 Protein;HEK293;200UG;CT4,CTLA4,CD152,CELIAC3,CTLA-4,GRD4,GSE,ICOS,IDDM12
Supplier:  ACROBiosystems CT4M52H5200UG
Encompass_Preferred
Recombinant Protein;Mouse CTLA-4 / CD152 Protein;200UG;Mouse CTLA-4, His Tag (CT4-M52H5) is expressed from human 293 cells (HEK293). It contains AA Glu 36 - Phe 162 (Accession # NP_033973).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.9 kDa. The protein migrates as 25-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse B7-1, Fc Tag (Cat. No. CD0-M5259) at 2 μg/mL (100 µl/well),can bind Mouse CTLA-4, His Tag (Cat. No. CT4-M52H5) with a linear range of 0.02-0.8 ng/mL (QC tested).;Immobilized Mouse B7-2, Fc Tag (Cat. No. CD6-M5251) at 5 μg/mL (100 μL/well) can bind Mouse CTLA-4, His Tag (Cat. No. CT4-M52H5) with a linear range of 0.4-13 ng/mL (Routinely tested).
Catalog No. 50-108-9559