Learn More
ACROBiosystems Recombinant Protein;Mouse SIRP alpha / CD172a Protein;HEK293;100UG;SIA,SIRP alpha,SHPS1,SIRPA,CD172A,BIT,SHPS-1,MFR,MYD-1,MYD1,PTPNS1,SIRPalpha,p84,SIRP-alpha,CD172
Supplier:  ACROBiosystems SIAM52H4100UG
Encompass_Preferred
Recombinant Protein;Mouse SIRP alpha / CD172a Protein;100UG;Mouse SIRP alpha (SIA-M52H4) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Asn 373 (Accession # P97797-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.8 kDa. The protein migrates as 60-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse CD47, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD7-M525a) at 2 μg/mL (100 μL/well) can bind Mouse SIRP alpha (Cat. No. SIA-M52H4) with a linear range of 0.02-0.625 μg/mL (QC tested).
Catalog No. 50-165-7603