Learn More
ACROBiosystems Recombinant Protein;Human HVEM / TNFRSF14 Protein, Fc Tag;HEK293;100UG;HVM,TNFRSF14,ATAR,HVEA,HVEM,LIGHTR,TR2,CD270
Supplier: ACROBiosystems HVMH5258100UG
Recombinant Protein;Human HVEM / TNFRSF14 Protein, Fc Tag;100UG;Human HVEM, Fc Tag (HVM-H5258) is expressed from human 293 cells (HEK293). It contains AA Leu 39 - Val 202 (Accession # NP_003811.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.4 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LIGHT, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. LIT-H5256) at 2 μg/mL (100 μL/well) can bind Human HVEM, Fc Tag (Cat. No. HVM-H5258) with a linear range of 0.3-20 ng/mL (QC tested).;Immobilized Human HVEM, Fc Tag (Cat. No. HVM-H5258) at 5 μg/mL (100 μL/well) can bind Biotinylated Human BTLA, Fc Tag (Cat. No. BTA-H82F8) with a linear range of 0.156-5 μg/mL (Routinely tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.