Learn More
ACROBiosystems Recombinant Protein;Human NTB-A / SLAMF6 Protein;HEK293;100UG;NTA,NTB-A,SLAMF6,Ly108,NK-T-B-antigen,CD352,KALI
Supplier:  ACROBiosystems NTAH52HA100UG
Encompass_Preferred
Recombinant Protein;Human NTB-A / SLAMF6 Protein;100UG;Human NTB-A (NTA-H52Ha) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Met 226 (Accession # Q96DU3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.0 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7825