Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CD23 / Fc epsilon RII Protein, His Tag & Avi Tag (Avitag™);HEK293;25UG;CD3,FCER2,FCER-2,CD23,CD-23,CD23A,CD-23A,CLEC4J,FCE2,FCE-2,IGEBF,FcεRII,FcεRⅡ,Fcr,Fcgr,Fc receptor,Fc epsilon receptor
Supplier: ACROBiosystems CD3H82Q525UG
Recombinant Protein;Biotinylated Human CD23 / Fc epsilon RII Protein, His Tag & Avi Tag (Avitag™);25UG;Biotinylated Human CD23, His Tag (CD3-H82Q5) is expressed from human 293 cells (HEK293). It contains AA Asp 48 - Ser 321 (Accession # P06734-1).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 34.2 kDa. The protein migrates as 40-46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Immunoglobulin E, Human Plasma at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD23, His Tag (Cat. No. CD3-H82Q5) with a linear range of 0.313-5 μg/mL (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.