Learn More
ACROBiosystems Recombinant Protein;Recombinant HCMV Glycoprotein B (gB), Fc Tag;HEK293;50UG;CMB,CMV,HCMV,GB,Envelope glycoprotein B,Glycoprotein GP55,UL55,GP55
Supplier:  ACROBiosystems CMBV525550UG
Encompass_Preferred
Recombinant Protein;Recombinant HCMV Glycoprotein B (gB), Fc Tag;50UG;Recombinant Human cytomegalovirus (HCMV) Glycoprotein B /gB Protein, C-Fc Tag (rh CMV-GB Fc Chimera) Val 23 - Lys 700 (Accession # AAA45920.1) with furin cleavage site mutated from 'RTKR' to 'TTQT', was produced in human 293 cells (HEK293) at ACROBiosystems.;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 104 kDa. The protein migrates as 140-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8613