Learn More
ACROBiosystems Recombinant Protein;Human BACE-1 Protein, Fc Tag;HEK293;200UG;BA1,BACE1,BACE-1,ASP2,BACE,FLJ90568,HSPC104,KIAA1149,memapsin-2,memapsin 2
Supplier: ACROBiosystems BA1H5261200UG
Recombinant Protein;Human BACE-1 Protein, Fc Tag;200UG;Human BACE-1, Fc Tag (BA1-H5261) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Thr 457 (Accession # NP_036236.1).;Human BACE-1, Fc Tag is fused with a human IgG1 Fc tag at the C-terminus, and has a calculated MW of 75 kDa. The predicted N-terminus is Glu. The reducing (R) protein migrates as 100-110 kDa in SDS-PAGE due to glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.