Learn More
ACROBiosystems Recombinant Protein;Human Activin RIIB / ACVR2B Protein;HEK293;200UG;ACB,ACVR2B,ACTRIIB,ACVR-2B,ACTR-IIB,ActR-IIB,ACTRⅡB,ACTR-ⅡB,MGC116908,ACVRIIB,Activin RIIB
Supplier:  ACROBiosystems ACBH5226200UG
Encompass_Preferred
Recombinant Protein;Human Activin RIIB / ACVR2B Protein;200UG;Human ACVR2B, His Tag (ACB-H5226) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Thr 137 (Accession # NP_001097).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.5 kDa. The protein migrates as 25-32 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-9075