Learn More
ACROBiosystems Recombinant Protein;Cynomolgus / Rhesus macaque B7-2 / CD86 Protein, Fc Tag;HEK293;100UG;CD6,CD86,CD-86,B7-2,B70,B-70,CD28LG2,LAB72,LAB-72,MGC34413
Supplier:  ACROBiosystems CD6C5254100UG
Encompass_Preferred
Recombinant Protein;Cynomolgus / Rhesus macaque B7-2 / CD86 Protein, Fc Tag;100UG;Cynomolgus / Rhesus macaque B7-2, Fc Tag (CD6-C5254) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Accession # G7NXR4).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52 kDa. The protein migrates as 80-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque CTLA-4, His Tag (Cat. No. CT4-C5227) at 2 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque B7-2, Fc Tag (Cat. No. CD6-C5254) with a linear range of 0.039-1.25 μg/mL (QC tested).
Catalog No. 50-165-9128
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.