Learn More
Sino Biological Recombinant Mouse FGFR4 Protein (His & hFc Tag)
Supplier:  Sino Biological 50194M03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Asp 366) of mouse FGFR4 (NP_032037.2) precursor was fused with the C-terminal polyhistidine tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1556