Learn More
ACROBiosystems ACROBiosystems Human Activin RIB / ACVR1B Protein, His Tag
Supplier: ACROBiosystems ACBH52H4100UG
Recombinant Protein;100UG;Human Activin RIB, His Tag (ACB-H52H4) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Glu 126 (Accession # P36896-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 13.3 kDa. The protein migrates as 16-22 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.