Learn More
ACROBiosystems ACROBiosystems Mouse CD24 Protein, His Tag
Supplier: ACROBiosystems CD4M52H7100UG
Recombinant Protein;100UG;Mouse CD24, His Tag (CD4-M52H7) is expressed from human 293 cells (HEK293). It contains AA Asn 27 - Gly 53 (Accession # P24807-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 4.6 kDa.