Learn More
ACROBiosystems ACROBiosystems Human MIS RII Protein, Fc Tag
Supplier: ACROBiosystems MIIH5258100UG
Recombinant Protein;100UG;Human MIS RII, Fc Tag (MII-H5258) is expressed from human 293 cells (HEK293). It contains AA Pro 18 - Ser 144 (Accession # Q16671-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.0 kDa.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.