Learn More
ACROBiosystems ACROBiosystems Human Siglec-8 Protein, Fc Tag
Supplier: ACROBiosystems SI8H5254100UG
Recombinant Protein;100UG;Human Siglec-8, Fc Tag (SI8-H5254) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Ala 363 (Accession # Q9NYZ4-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.3 kDa. The protein migrates as 70-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.