Learn More
ACROBiosystems ACROBiosystems Human Mucin-1 / MUC-1 Protein, Llama IgG2b Fc Tag, low endotoxin
Supplier: ACROBiosystems MU1H5255100UG
Recombinant Protein;100UG;Human Mucin-1, Llama IgG2b Fc Tag, low endotoxin (MU1-H5255) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Gly 167 (Accession # AAI20976).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 42.4 kDa. The protein migrates as 50-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.