Learn More
ACROBiosystems ACROBiosystems Canine M-CSF R / CSF1R / CD115 Protein, His Tag
Supplier: ACROBiosystems CSRC82H3100UG
Recombinant Protein;100UG;Canine M-CSF R, His Tag (CSR-C82H3) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 510 (Accession # E2RJV2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 56.1 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Canine M-CSF R, His Tag (Cat. No. CSR-C82H3) at 2 μg/mL (100 μL/well) can bind Biotinylated Human M-CSF, His Tag, Avi Tag (Cat. No. MCF-H82E6) with a linear range of 1-16 ng/mL (QC tested).
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.