Learn More
ACROBiosystems ACROBiosystems Human CD19 (20-291) Protein, Mouse IgG2a Fc Tag
Supplier: ACROBiosystems CD9H5258100UG
Recombinant Protein;100UG;Human CD19 (20-291), Mouse IgG2a Fc Tag (CD9-H5258) is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 291 (Accession # P15391-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 57.0 kDa. The protein migrates as 75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD19 (20-291), Mouse IgG2a Fc Tag (Cat. No. CD9-H5258) at 5 μg/mL (100 μL/well) can bind Mouse / Human chimeric FMC63 MAb (human IgG1) with a linear range of 0.5-8 ng/mL (QC tested).