Learn More
ACROBiosystems ACROBiosystems Human GM-CSF R alpha Protein, Fc Tag
Supplier:  ACROBiosystems GRAH5255100UG
Encompass_Preferred
Recombinant Protein;100UG;Human GM-CSF R alpha, Fc Tag (GRA-H5255) is expressed from human 293 cells (HEK293). It contains AA Glu 23 - Gly 320 (Accession # P15509-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.9 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GM-CSF, Tag Free (Cat. No. GMF-H4214) at 1 μg/mL (100 μL/well) can bind Human GM-CSF R alpha, Fc Tag (Cat. No. GRA-H5255) with a linear range of 0.5-16 ng/mL (QC tested).
Catalog No. 50-195-7080