Learn More
NEOGEN Reveal 2.0 for Salmonella
Supplier: NEOGEN 700002795
The Reveal 2.0 for Salmonella test provides for the rapid recovery of Salmonella in chicken carcass rinse, raw ground turkey, raw ground beef, hot dogs, raw shrimp, ready-to-eat meal products, dry pet food, ice cream, fresh spinach, cantaloupe, peanut butter, swabs from stainless steel surfaces, and sprout irrigation water samples allowing detection and presumptive identification of Salmonella within 24 hours.Approvals: AOAC-RI 960801Limit of Detection: 1 cfu/analytical unitTesting Time: 15 MinutesTime to Result: 24-48 HoursFor more information visit https://www.neogen.com/solutions/microbiology/reveal-2-salmonella/