Learn More
ACROBiosystems Human CD40 Ligand / TNFSF5 Protein, Fc Tag (active trimer) (MALS verified)
Supplier: ACROBiosystems CDLH5269100UG
Recombinant Protein;100UG;Human CD40 Ligand, Fc Tag (active trimer) (MALS verified) (CDL-H5269) is expressed from human 293 cells (HEK293). It contains AA Gly 116 - Leu 261 (Accession # P29965-1) trimer Design.;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 74.7 kDa. The protein migrates as 120-150 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD40, His Tag (Cat. No. CD0-H5228) at 1 μg/mL (100 μL/well) can bind Human CD40 Ligand, Fc Tag (active trimer) (MALS verified) (Cat. No. CDL-H5269) with a linear range of 0.2-3 ng/mL (QC tested).;FACS analysis shows that Human CD40 Ligand, Fc Tag (active trimer) (MALS verified) (Cat. No. CDL-H5269) can bind to 293T-CD40 cells surface CD40. The concentration of Human CD40 Ligand is 0.3 μg/mL (Routinely tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.