Learn More
ACROBiosystems Human CD27 / TNFRSF7 Protein, Fc Tag
Supplier:  ACROBiosystems CD7H5254200UG
Encompass_Preferred
Recombinant Protein;200UG;Human CD27, Fc Tag (CD7-H5254) is expressed from human 293 cells (HEK293). It contains AA Thr 21 - Ile 192 (Accession # NP_001233).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.9 kDa. The protein migrates as 53-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD27, Fc Tag (Cat. No. CD7-H5254) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD27 Ligand, Fc Tag (Cat. No. TN7-H82F4) with a linear range of 1-31 ng/mL (QC tested).;Flow Cytometry assay shows that Human CD27, Fc Tag (Cat. No. CD7-H5254) can bind to Raji cells Surface Human CD70. The concentration of CD27 used is 1 μg/mL (Routinely tested).
Catalog No. 50-201-9875