Learn More
ACROBiosystems Biotinylated Mouse IL-6 R alpha / CD126 Protein, His,Avitag™
Supplier:  ACROBiosystems ILRM82E925UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Mouse IL-6 R alpha, His,Avitag (ILR-M82E9) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Glu 357 (Accession # P22272-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 41.0 kDa. The protein migrates as 45-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse Human IL-6, His Tag (Cat. No. IL6-M5245) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse IL-6 R alpha, His,Avitag (Cat. No. ILR-M82E9) with a linear range of 20-156 ng/mL (QC tested).
Catalog No. 50-205-5137
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.