Learn More
ACROBiosystems FITC-Labeled Human CD38 Protein, Fc Tag
Supplier:  ACROBiosystems CD8HF25525UG
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human CD38, Fc Tag (Cat. No. CD8-HF255) is expressed from human HEK293 cells. It contains AA Val 43 - Ile 300 (Accession # P28907-1). It is the FITC labeled form of Human CD38 Protein, Fc Tag (Cat. No. CD8-H5255).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 56.3 kDa. The protein migrates as 65-75 under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-CD38 Antibody (Daratumumab), Human IgG1 at 2 μg/mL (100 μL/well) can bind FITC-Labeled Human CD38, Fc Tag (Cat. No. CD8-HF255) with a linear range of 0.078-0.625 μg/mL (QC tested).;Immobilized Monoclonal Anti-CD38 Antibody, Human IgG1 at 2 μg/mL (100 μL/well) can bind FITC-Labeled Human CD38, Fc Tag (Cat. No. CD8-HF255) with a linear range of 0.039-1.25 ug/mL (Routinely tested).
Catalog No. 50-205-5176