Learn More
ACROBiosystems Biotinylated Human NKG2A&CD94 Protein, Fc,Avitag™
Supplier: ACROBiosystems NC4H82F525UG
Recombinant Protein;25UG;Biotinylated Human NKG2A&CD94, Fc,Avitag™ (NC4-H82F5) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Leu 233 & Asp 57-Ile 179 (Accession # P26715-1 (NKG2A) & Q13241-1 (CD94)).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 40.4 kDa & 42.3 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human NKG2A&CD94, Fc,Avitag (Cat. No. NC4-H82F5) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate. can bind various dilution ratio of HLA-E*01:03 HLA-A leader3-11 Tetramer-VMAPRTLVL-PE (QC tested).