Learn More
ACROBiosystems Human BCMA 2.5mg
Supplier: ACROBiosystems MBSC0042.5MG
ActiveMax Human BCMA uBeads premium grade (for cells) are uniform superparamagnetic beads of 5.5 µm with streptavidin on its surface and coupled with biotinylated Human BCMA Protein expressed from human 293 cells (HEK293) and contains AA Met 1 - Ala 54 (Accession # Q02223-1).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.