Learn More
ACROBiosystems Human IL-3 R alpha 25ug
Supplier:  ACROBiosystems ILAH821425UG
Encompass_Preferred
Biotinylated Human IL-3 R alpha Protein Avitag (ILA-H8214) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Arg 305 (Accession # P26951-1).
Catalog No. 50-253-4601