Learn More
ACROBiosystems Human DLL4 200ug
Supplier:  ACROBiosystems GMPDL4H23200UG
Encompass_Preferred
GMP Biotinylated Human DLL4 Protein HisAvitag (GMP-DL4H23) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Pro 524 (Accession # NP_061947.1).
Catalog No. 50-253-4993