Learn More
ORS Nasco T5900 REGULAR BK FR CLR HS
Supplier:  ORS Nasco 763T5900LTKHS
Encompass_Preferred
Honeywell part# T5900LTKHS. Honeywell North Adaptec Uvex HydroShield Anti-Fog Coating Safety Glasses Description: Regular Size Black/Red Frame Clear Lens Frame Color: Black/Red Frame Material: Polycarbonate Frame Type: Regular Lens Tint: Clear Lens Type: Anti-fog Anti-static Scratch-resistant Safety glasses
Catalog No. 50-255-1565