Learn More
ACROBiosystems Biotinylated Mouse H-2Db&B2M Monomer Protein (Peptide free, MALS verified)
Supplier:  ACROBiosystems H2MM82E925UG
Encompass_Preferred
Biotinylated Mouse H-2Db&B2M Monomer Protein (H2M-M82E9) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Val 309 (H-2Db) & Ile 21 - Met 119 (B2M) (Accession # P01899 (H-2Db) & P01887 (B2M)).
Catalog No. 50-271-9523