Learn More
ACROBiosystems GMP ActiveMax Human T cell Activation/Expansion CD3/CD28 Beads
Supplier: ACROBiosystems GMPMBS00110MG
GMP ActiveMax Human T cell Activation/Expansion CD3/CD28 Beads are uniform 5.5 μm of magnetic beads coated with an optimized mixture of GMP grade mouse monoclonal antibodies against the CD3 and CD28, mimicking in vivo stimulation by APCs.GMP ActiveMax Human T cell Activation/Expansion CD3/CD28 Beads are Manufactured under a GMP compliance factory with animal free raw materials, and tested under GMP guidelines.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.