Learn More
Med Vet International McKesson Sanitizing Skin Wipe, Unscented, 269, 100/Box
Supplier:  Med Vet International 865276BX
Encompass_Preferred
Sanitizing Skin Wipe McKesson Individual Packet BZK (Benzalkonium Chloride) Unscented 100 Count Designed for general antiseptic cleansing for both patients and clinicians Each towelette contains 0.13% w/v benzalkonium chloride to help disinfect Sterile towelettes are 5 x 7 inches
Catalog No. 50-304-5162