Learn More
Sino Biological Recombinant Mouse kynureninase/KYNU Protein (His Tag)
Supplier:  Sino Biological 50310M08B100
Encompass_Preferred
A DNA sequence encoding the mouse KYNU (Q9CXF0) (Met 1-Ser 464) was expressed,with a C-terminal polyhistidine tag.
Catalog No. 50-162-1720