Learn More
COLDER PRODUCTS COMPANY FEMALE INSERT 1/4 ID HB 25/PK
Supplier: COLDER PRODUCTS COMPANY PLCD17004
CPC Plastic Connectors Couplings and Fittings Buna-N Seal Description 1 8 L x 0 94 in H (45 7 x 23 mm) Hose Barb Valved In-line Coupling Body White For Use With 6 4mm Includes Plastic connector Overall Length 1 8 in 45 7 mm Height 0 94 in 23 mm Maximum Pressure 120 psi