Learn More
Sino Biological Recombinant Mouse ICAM-1/CD54 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50440M08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse ICAM1 (NP_034623.1) (Met 1-Asn 485) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1910