Learn More
Sino Biological Recombinant Mouse CHODL Protein (His Tag)
Supplier:  Sino Biological 50699M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CHODL (EDK98299.1) (Met 1-Asn 216) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2350