Learn More
Sino Biological Recombinant Mouse CLEC10A/CD301 Protein (His Tag)
Supplier:  Sino Biological 50707M07H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CLEC10A (NP_001191181.1) (Gln 58-Ser 305) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-2367