Learn More
Sino Biological Recombinant Mouse CD45 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 50713M02H1100
Encompass_Preferred
A DNA sequence encoding the mouse PTPRC (NP_035340.3) (Met1-Leu429) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2392